Learn More
Medchemexpress LLC Mono-(6-p-toluenesulfonyl)-β-cyclodextrin | 67217-55-4 | MFCD05864975 | 95.0% | 1289.17 | C49H76O37S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW129395100MG
Encompass_Preferred
Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a tosylated derivative of β-cyclodextrin supplied as a white to off-white solid for use as a biochemical reagent and synthetic intermediate. It introduces a reactive tosyl group at the 6-position for further functionalization and is commonly used in host-guest studies and inclusion complex research. Typical purity is 95.0%.
  • White to off-white solid form.
  • Tosylated at the 6-position enabling nucleophilic substitution.
  • Useful building block for cyclodextrin functionalization.
  • Suitable for host-guest and inclusion complex studies.
  • Typical purity 95.0% and multiple pack sizes available.
Catalog No. 50-002-83552