Learn More
Apexbio Technology LLC Monoacetylphloroglucinol 480-66-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC C30865.1
Encompass_Preferred
Monoacetylphloroglucinol (CAS 480-66-0) is a phenolic small-molecule metabolite produced by certain Pseudomonas strains It serves as an acetylated intermediate substrate in the biosynthesis of 2 4-diacetylphloroglucinol via MAPG acetyltransferase-mediated catalysis Monoacetylphloroglucinol exhibits broad-spectrum antibiotic activity though its potency is limited as shown by relatively high antibacterial IC50 values in preliminary in vitro studies Key experimental models and precise IC50 metrics are not specified Based on these pharmacological properties Monoacetylphloroglucinol is primarily researched for its potential in microbial interactions against phytopathogenic organisms and the development of agricultural biocontrol strategies
Catalog No. 50-000-87909