Learn More
Selleck Chemical LLC Monobenzone 50mg 103-16-2
SDP
Supplier:  Selleck Chemical LLC S165250MG
Encompass_Preferred
Monobenzone is a compound used as a topical agent for medical depigmentation. It can specifically block tyrosinase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-596-8