Learn More
Medchemexpress LLC HY-N7143 500mg Medchemexpress, Monobutyl phthalate CAS:131-70-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7143500MG
Encompass_Preferred
Medchemexpress, HY-N7143 500mg Monobutyl phthalate CAS:131-70-4 Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP), possesses antiandrogenic effects. Monobutyl phthalate is an embryotoxicant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2987