Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monobutyl phthalate-d4 | 478954-81-3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN7143S5MG

Encompass_Preferred

Monobutyl phthalate-d4 is a deuterium-labeled form of Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP). It is known for its antiandrogenic effects and is considered an embryotoxicant.

  • High purity of 99.33% (HPLC)
  • Isotopic enrichment of 99.3%
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular formula: C12H10D4O4
  • Molecular weight: 226.26
  • Appearance: White to off-white solid

Catalog No. 50-004-17969


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.