Learn More
Medchemexpress LLC Monobutyl phthalate (Standard) | 131-70-4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN7143R50MG
Encompass_Preferred
Monobutyl phthalate (Standard) is the analytical standard of Monobutyl phthalate, intended for research and analytical applications. This compound is a major metabolite of dibutyl phthalate (DBP) and possesses antiandrogenic effects, also being an embryotoxicant.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-004-19877