Learn More
Medchemexpress LLC Monocalcium glycerophosphate | 27214-00-2 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB22031G
Encompass_Preferred
Monocalcium glycerophosphate is an inhibitor of intestinal alkaline phosphatase F3 and exhibits anti-cavity properties. It serves as a source of calcium and phosphorus in total parenteral nutrition solutions. It helps prevent the mineralization and development of bones from intravenous nutrition and contributes to maintaining the integrity of the intestinal epithelium.
  • Protects trans-epithelial electrical resistance (TEER) in Caco-2 cells and reduces the increase in mannitol permeability rate
  • Significantly decreases the level of demineralization in an in vitro bacterial flow chamber model
  • Acts as an effective source of calcium and phosphorus in piglet TPN solutions
  • Increased the retention of calcium and phosphorus, and bone mineralization improved in animal models
Catalog No. 50-004-12348