Learn More
ACROBiosystems Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Y45), 250 tests
Supplier:  ACROBiosystems FM3Y45250TESTS
Encompass_Preferred
FMC63 is an IgG2a mouse monoclonal antibody specific for CD19, which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. So far, most of reported CART19 trials contain the anti-CD19 scFv derived from FMC63, including the two FDA-approved CARs Kymriah and Yescarta.
Catalog No. NC1838164