Learn More
Quidel Corporation MONOCLONAL ANTI BODY HUMAN 10ul
SDP
Supplier:  Quidel Corporation A201
Encompass
A murine monoclonal antibody to an epitope on the globular head domain of C1q.
Catalog No. NC1720269