Learn More
ATCC Monoclonal Antibody;Anti-SARS-CoV-2 Receptor; Binding Domain (RBD);protein, Clone 2TP1E6
Supplier:  ATCC VR3332
Encompass
Monoclonal antibody prepared against the Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) spike glycoprotein receptor binding domain. This product was purified from clone 2TP1E6 hybridoma supernatant by protein L affinity chromatography.
Catalog No. 50-238-2469