Learn More
ATCC Monoclonal Antibody;Anti-SARS-CoV-2 Receptor; Binding Domain (RBD);protein, Clone 2TP2E7
Supplier:  ATCC VR3336
Encompass
Monoclonal antibody prepared against the Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) spike glycoprotein receptor binding domain. This product was purified from clone 2TP2E7 hybridoma supernatant by protein G affinity chromatography.
Catalog No. 50-238-3341