Learn More
Selleck Chemical LLC Monocrotaline 25mg 315-22-0 Crotaline
SDP
Supplier:  Selleck Chemical LLC S381225MG
Encompass_Preferred
Monocrotaline (MCT, Crotaline) is an 11-membered macrocyclic pyrrolizidine alkaloid (PA) that causes a pulmonary vascular syndrome in rats characterized by proliferative pulmonary vasculitis, pulmonary hypertension, and cor pulmonale. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8267