Learn More
Medchemexpress LLC Monocrotaline | 315-22-0 | 325.36 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYN0750200MG
Encompass_Preferred
Monocrotaline is an 11-membered macrocyclic pyrrolizidine alkaloid with antitumor activity. It inhibits OCT-1 and OCT-2 and is cytotoxic to hepatocellular carcinoma cells. This compound is also used to induce a model of pulmonary hypertension in rodents. It exhibits dose-dependent cytotoxicity and genotoxicity against HepG2 cells in vitro.
  • Inhibits OCT-1 and OCT-2.
  • Exhibits antitumor activity.
  • Cytotoxic to hepatocellular carcinoma cells.
  • Used to induce models of pulmonary hypertension in rodents.
  • Shows dose-dependent cytotoxicity and genotoxicity against HepG2 cells.
Catalog No. 50-004-15298