Learn More
Medchemexpress LLC Monogalactosyldiacylglycerol | 1932659-76-1 | 775.06 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN125405MG
Encompass_Preferred
Monogalactosyldiacylglycerol (MGDG) is a polar lipid found in the chloroplast thylakoid of aerobic photosynthetic organisms. It does not form a bilayer and lacks charge, which creates a fluid environment facilitating electron transfer in photosynthesis. It may also be utilized in the packaging of natural proteins and exhibits anti-inflammatory effects in vivo.
  • Polar lipid
  • Does not form a bilayer
  • Found in chloroplast thylakoid of aerobic photosynthetic organisms
  • Lacks charge
  • Provides a fluid environment for electron transfer in photosynthesis
  • May be used in the packaging of natural proteins
  • Has anti-inflammatory effects in vivo
Catalog No. 50-004-15040