Learn More
Medchemexpress LLC Monoisodecyl phthalate-d4 | 1398065-94-5 | MFCD29045533 | >95.0% | 310.42 g/mol | C18H22D4O4 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYW6542782.5MG
Encompass_Preferred
Monoisodecyl phthalate-d4 is a deuterium-labeled internal standard of 1-(8-methylnonyl) 1,2-benzenedicarboxylate used as a tracer and quantitative internal standard for NMR, GC-MS, and LC-MS analyses. The deuterated analog improves mass spectrometric discrimination and supports accurate quantitation in environmental, food, and bioanalytical workflows. Certificate of Analysis provides exact purity and storage instructions.
  • Deuterium-labeled internal standard for accurate quantitation.
  • Compatible with NMR, GC-MS, and LC-MS analytical workflows.
  • Molecular formula C18H22D4O4; molecular weight 310.42 g/mol.
  • Designed for use as a tracer and internal standard in environmental and bioanalytical studies.
  • Certificate of Analysis available for purity and storage details.
Catalog No. 50-002-84956