Learn More
Echelon Biosciences Research Labs MONOLYSOCARDIOLIPIN 16:0 100ug
SDP
Supplier:  Echelon Biosciences Research Labs LM160100UG
Encompass
Cardiolipin (CL) is an important component inprokaryotic and in eukaryotic inner mitochondrial membranes. In humans, CL is made by cardiolipin synthase from phosphatidylglycerol and cytidinediphosphate-diacylglycerol. Monolysocardiolipin is an intermediate in CL remodeling, and acetyl transferase tafazzin mutations impairing linoleic acid transfer to MCL are considered the main reason of Barth syndrome. Autoantibodies to CL have been found in patients with anti-phospholipid syndrome and lupus. Trispalmitoyl MCL is a stable, synthetic analog of this rare and interesting phospholipid. This item is non returnable
Catalog No. 50-822-29