Learn More
ILEX LIFE SCIENCES LLC MONOMERIC CRP ELISA 96 WELLS
SDP
Supplier:  ILEX LIFE SCIENCES LLC PL1026
Encompass_Preferred
PromedeusLab Monomeric CRP (mCRP) Human ELISA Kit (96 wells). DESCRIPTION: Enzyme immunoassay for the quantitative determination of Monomeric C-Reactive Protein (mCRP) in human serum and plasma samples. Quality controls are included with the kit. Developed and manufactured in Europe by PromedeusLab. SAMPLE TYPES: Plasma or serum. ASSAY RANGE: 1.25 ng/mL - 80 ng/mL. SENSITIVITY: The limit of detection, defined as a concentration of human MCRP giving absorbance higher than absorbance of blank + 3 standard deviations, is better than 0.63 ng/mL of sample. RESEARCH AREA: Deposition of mCRP, which has a much lower aqueous solubility than pCRP, has been shown in the brain in infarcted areas of Alzheimer's Disease patients and in regions with amyloid burden, in atherosclerotic plaques in vascular disease and in other foci of inflammatory tissue injuries.
Catalog No. 50-273-3631