Learn More
Medchemexpress LLC HY-15162 10mg Medchemexpress, Monomethyl auristatin E CAS:474645-27-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1516210MG
Encompass_Preferred
Medchemexpress, HY-15162 10mg Monomethyl auristatin E CAS:474645-27-7 Monomethyl auristatin E (MMAE; SGD-1010) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by inhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9087