Learn More
Medchemexpress LLC Monomethyl auristatin E | 474645-27-7 | MFCD09935109 | C68H105N9O15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15162G100MG
Encompass_Preferred
Monomethyl auristatin E (MMAE) (GMP) is a tubulin polymerization inhibitor produced using GMP guidelines.
  • Produced using GMP guidelines
  • Tubulin polymerization inhibitor
  • Efficiently released from SGN-35 within CD30+ cancer cells
  • Able to exert cytotoxic activity on bystander cells due to membrane permeability
  • Sensitizes colorectal and pancreatic cancer cells to IR in a schedule and dose-dependent manner correlating with mitotic arrest
  • Radiosensitization evidenced by decreased clonogenic survival and increased DNA double-strand breaks in irradiated cells
  • In combination with IR, results in tumor growth delay
  • Tumor-targeted ACPP-cRGD-MMAE with IR produces a more robust and significantly prolongs tumor regression in xenograft models
Catalog No. 50-003-72900