Learn More
Medchemexpress LLC Monomethyl auristatin E-SMCC conjugate | 2021179-11-1 | 95.7% | 1140.43 g/mol | C58H89N7O14S | 1 MG

Supplier: Medchemexpress LLC HY1356601MG
MMAE-SMCC is a drug-linker conjugate used to prepare antibody-drug conjugates (ADCs) for research. It couples monomethyl auristatin E (MMAE), a potent mitotic and tubulin inhibitor, to an SMCC linker to enable covalent attachment to antibodies. Supplied as a dry powder, the compound is intended for research use only and is not for human or clinical use.
- Composed of monomethyl auristatin E (MMAE) and an SMCC linker.
- Designed for antibody-drug conjugate (ADC) research use.
- Purity approximately 95.7%.
- Molecular weight 1,140.43 g/mol; chemical formula C58H89N7O14S.
- Available in mg-scale packages (1 mg, 5 mg, 10 mg; larger quantities on request).
- Store at -20°C, protect from light; unstable in solution, prepare fresh.
- Not for human or clinical use; for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.