Learn More
Medchemexpress LLC Monomethylauristatin F tert-butyl ester | 745017-95-2 | 99.8% | 788.07 | C43H73N5O8 | 50 MG

Supplier: Medchemexpress LLC HY15622550MG
MMAF-OtBu is a tert-butyl protected derivative of monomethylauristatin F intended for use as a cytotoxic payload in antibody-drug conjugate (ADC) synthesis. It is supplied as a solid or as a DMSO solution for research use and is accompanied by supporting documents for characterization and safe handling.
- High purity: 99.83% (analytical)
- Molecular weight: 788.07
- Chemical formula: C43H73N5O8
- Available in multiple pack sizes, including 50 MG
- Suitable for ADC synthesis as a cytotoxic payload
- Documents available: data sheet, COA, SDS, handling instructions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.