Learn More
Medchemexpress LLC Monomethylauristatin F tert-butyl ester (MMAF-OtBu) | 745017-95-2 | 99.8% | 788.07 | C43H73N5O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1562255MG
Encompass_Preferred
MMAF-OtBu is a tert-butyl ester derivative of monomethylauristatin F used as a cytotoxic payload in antibody-drug conjugate (ADC) synthesis for research applications.
  • High purity compound for research use only.
  • Potent tubulin polymerization inhibitor, used as ADC payload.
  • Molecular formula C43H73N5O8; molecular weight 788.07 g/mol.
  • Provided in small-mass packages suitable for synthetic chemistry (e.g., 5 mg).
  • Store solid at -20°C; in solution keep at -80°C for long-term storage.
Catalog No. 50-002-04391