Learn More
Medchemexpress LLC Monopropyl phthalate | 4376-19-6 | >98.0% | 208.21 g/mol | C11H12O4 | 50 MG

Supplier: Medchemexpress LLC HYW70339850MG
Monopropyl phthalate is a phthalate monoester and a metabolic hydrolysis product of dipropyl phthalate, supplied as a high-purity small-molecule standard for analytical chemistry, toxicology, and environmental studies.
- High purity: >98.0%.
- Packaged as a 50 mg standard suitable for analytical assays.
- Intended for metabolite identification and method development.
- Molecular weight 208.21 g/mol; formula C11H12O4.
- CAS number 4376-19-6 for reliable substance identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.