Learn More
Genscript Corporation MonoRab MSH6 3B6 mAb Rab
Supplier:  Genscript Corporation V0820110
Encompass
MSH6 also known as MutS protein homolog 6 is a member of the Mutator S (MutS) family of proteins It functions to repair mutations during DNA replication MSH6 is a useful in diagnostics of gastrointestinal tract cancer
Catalog No. 50-000-58789
Only null left
Add to Cart
Request a Quote