Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Cocktail [HRP]
Supplier:  Genscript Corporation A02016200
Encompass
GenScript MonoRab™ Rabbit Anti-Camelid VHH Cocktail [HRP] is specific for Camelid sdAb (Llama and Camel). It has no cross-reactivity with mouse, rat, rabbit, goat or human immunoglobulins.
Catalog No. NC2126154