Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Cocktail [PE]
Supplier:  Genscript Corporation A02018200
Encompass
GenScript MonoRab™ Rabbit Anti-Camelid VHH Cocktail [PE] is specific for Camelid VHH antibody (Llama, Alpaca and Camel). It has no cross-reactivity with mouse, rat, rabbit, goat or human immunoglobulins.
Catalog No. NC1979778