Learn More
Medchemexpress LLC Monosodium 2-sulfoterephthalate | 19089-60-2 | MFCD00059843 | 99.0% | 269.19 g/mol | C8H6NaO7S | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW090309
Encompass_Preferred
Monosodium 2-sulfoterephthalate is a research-grade small molecule reported as a glutamate carboxypeptidase II inhibitor. It is supplied as a solid and as a DMSO solution for laboratory research use only. Typical specifications include CAS 19089-60-2, molecular formula C8H6NaO7S, molecular weight 269.19 g/mol, and high purity suitable for biochemical studies.
  • Glutamate carboxypeptidase II inhibitor activity
  • Available as solid and 10 mM in DMSO solution
  • Common pack sizes include 1 g, 5 g, 10 g, 25 g, 50 g
  • High purity suitable for research (approx. 99.0%)
  • Solid appearance for handling and storage
Catalog No. 50-002-09484