Learn More
Selleck Chemical LLC Monosodium L-glutamate hydrate
SDP
Supplier:  Selleck Chemical LLC E01525G
Encompass_Preferred
L( )-Monosodium glutamate monohydrate (MSG monohydrate L-Glutamic acid monosodium salt monohydrate Monosodium L-glutamate monohydrate Monosodium L-glutamate monohydrate) is a widespread nutritional additive and flavoring agent L( )-Monosodium glutamate monohydrate can lead to oxidative stress-mediated DNA damage and apoptosis L( )-Monosodium glutamate monohydrate can be used to induce animal models of Obesity
Catalog No. 50-313-6793