Learn More
Sigma Aldrich Fine Chemicals Biosciences Montana I soil NISTR SRMR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST2710A
Encompass
This Standard Reference Material (SRM) is intended primarily for use in the analysis of soils sediments or other materials of a similar matrix. One unit of SRM 2710a consists of 50 g of the dried powdered soil blended with lead oxide. For more information please refer to the COA and SDS.SRM 2710A cert SRM 2710A SDS
Catalog No. 50-290-7507