Learn More
TARGETMOL CHEMICALS INC MONTELUKAST SODIUM 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1677L500MG
Encompass_Preferred
Also available in 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Montelukast sodium (MK0476) is an orally available leukotriene receptor antagonist which is widely used for the prophylaxis and chronic treatment of asthma and has been linked to rare cases of clinically apparent liver injury. purity: 99%
Catalog No. 50-257-7326