Learn More
Medchemexpress LLC Monuron | 150-68-5 | 100.0% | 50 MG

Supplier: Medchemexpress LLC HYB187550MG
Monuron is a phenylurea herbicide that functions by inhibiting photosynthesis.
- It is a phenylurea herbicide.
- It inhibits photosynthesis.
- Available as a white to light yellow solid.
- High purity of 99.96%.
- Soluble in DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.