Learn More
Medchemexpress LLC Monuron | 150-68-5 | 99.96% | 1 ML

Supplier: Medchemexpress LLC HYB187510MM/1ML
Monuron is a phenylurea herbicide which inhibits photosynthesis. It is for research use only and not sold to patients.
- Purity is 99.96%
- Appearance is solid
- Color is white to light yellow
- Ships at room temperature in continental US
- Store as powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.