Learn More
Alkali Scientific MOPS [3 (N morpholino) propanesulfonic acid], 10 Kg
SDP
Supplier:  Alkali Scientific MOP003CP
Encompass_Preferred
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a biochemical reagent used for pH buffering in a variety of laboratory applications. This 10kg package of MOPS is ideal for large-scale production of buffer solutions, making it suitable for high-volume research and industrial applications. MOPS buffers are particularly effective in maintaining a stable pH range of 6.5 to 7.9, which is critical for biological assays, enzyme reactions, and protein analysis. It is commonly used in molecular biology, electrophoresis, and chromatography to stabilize proteins, nucleic acids, and enzymes. This bulk packaging ensures a cost-effective solution for labs with high buffer consumption.
Catalog No. 50-006-61636