Learn More
Alkali Scientific MOPS, Sodium Salt [3 (N Morpholino) propanesulfonic acid sodium salt], 100 G
SDP
Supplier:  Alkali Scientific MOP009CP
Encompass_Preferred
MOPS (3-(N-Morpholino)propanesulfonic acid sodium salt) is a widely used biochemical reagent for pH buffering in biological and biochemical applications. This 100g volume of MOPS sodium salt is ideal for maintaining stable pH levels in systems that require a buffered environment, particularly in protein and nucleic acid studies. Its stability at a pH range of 7.0–7.5 makes it ideal for enzyme reactions, cell cultures, and electrophoresis. MOPS is commonly used in molecular biology and biochemistry for experiments that require consistent pH regulation over extended periods. It is used in a variety of applications, including gel electrophoresis, enzyme assays, and protein purification protocols.
Catalog No. 50-006-60063