Learn More
Medchemexpress LLC MOPS (sodium salt) | 71119-22-7 | 99.8% | 50 G
SDP
Supplier:  Medchemexpress LLC HYD0859A50G
Encompass_Preferred
MOPS sodium salt is a buffering agent used in biological applications. It helps maintain the pH of mammalian cell culture media and significantly improves the thermal stability of BSA. This product is also suitable for the preparation of keratinocyte and microbial culture media.
  • Maintains pH of mammalian cell culture media
  • Enhances thermal stability of BSA
  • Suitable for preparing keratinocyte culture medium
  • Suitable for preparing microbial culture medium
  • Can be used in co-culture of keratinocytes and microorganisms
Catalog No. 50-004-10408