Learn More
TARGETMOL CHEMICALS INC MORACIN C 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN19495MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Moracin C a natural compound found in Morus mesozygia Morus alba var. multicaulis is an anti-inflammatory agent. moracin C inhibits nitric oxide (NO) release and LPS-activated reactive oxygen species (ROS) in cells. purity: 99%
Catalog No. 50-257-8887