Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moracin M | 56317-21-6 | 99.0% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY12294250MG

Encompass_Preferred

Moracin M is a phenolic metabolite isolated from Mori Cortex, supplied for research use. It shows reported phosphodiesterase-4 (PDE4) inhibitory activity and anti-inflammatory effects in cellular assays. Offered in solid powder and solution formats for biochemical and pharmacological research.

  • Phenolic metabolite with reported PDE4 inhibitory activity
  • High purity suitable for research use
  • Available as solid powder and 10 mM DMSO solution
  • Characterized chemical identifiers: CAS 56317-21-6, MFCD17166969
  • Molecular formula C14H10O4 and molecular weight 242.23 g/mol

Catalog No. 50-002-30683


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.