Learn More
Sigma Aldrich Fine Chemicals Biosciences MorDalphos Pd G35G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 7923575G
Encompass
MorDalphos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in SOLUTIONons. MorDalphos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings shorter reaction time efficient formation of the active catalytic species and accurate control of ligand palladium ratio.
Catalog No. 50-290-7233