Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Morellic acid | 5304-71-2 | 99.8% | 560.63 | C33H36O8 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN409410MG

Encompass_Preferred

Morellic acid is a naturally occurring compound isolated from Garcinia morella with reported antiangiogenic activity. It is provided as an analytical/research standard intended for laboratory research use only.

  • Natural product isolated from Garcinia morella with antiangiogenic activity.
  • Analytical/research standard suitable for in vitro and in vivo studies.
  • High purity: 99.81%.
  • Chemical identifiers: CAS 5304-71-2; molecular formula C33H36O8; molecular weight 560.63.
  • Solubility: DMSO 100 mg/mL (in vitro); in vivo formulations ≥ 2.5 mg/mL (clear solutions).
  • Recommended storage: 4°C for powder; in solvent -80°C (6 months) or -20°C (1 month), sealed away from moisture and light.
  • Available in small pack sizes suitable for compound screening and pharmacology.

Catalog No. 50-002-89852


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.