Learn More
Cayman Chemical Mornteltartrate 1g
SDP
Supplier:  Cayman Chemical 229021 G
Encompass_Preferred
A positive allosteric modulator of nAChRs; enhances channel gating of the α3β2 nAChR subtype
Catalog No. 50-299-4048