Learn More
Medchemexpress LLC Moroxydine hydrochloride | 3160-91-6 | 99.4% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0420A500MG
Encompass_Preferred
Moroxydine hydrochloride, also known as ABOB hydrochloride, is a broad-spectrum antiviral agent effective against various DNA and RNA viruses such as influenza, herpes simplex, measles, mumps, and hepatitis C. It shows excellent antiviral activity and low cytotoxicity against dsRNA and large DNA viruses.
  • Protects ctenopharyngodon idella kidney (CIK) and grass carp ovary (GCO) cells from GCRV.
  • Protects epithelioma papulosum cyprinid (EPC) cells from giant salamander iridovirus (GSIV).
  • Blocks virus-induced cytopathic effects and cell death.
  • Decreases apoptosis in GCRV and GSIV infected cells.
  • Inhibits caspase 3 activity, Bax expression, and down-regulates Bcl-2.
  • Suppresses viral gene expression.
  • Arrests cell cycle in G1/S and G2/M phases.
  • Inhibits GCRV-induced upregulation of immune genes (IL-1β, IFN-I, TNF-α).
Catalog No. 50-004-07042