Learn More
Medchemexpress LLC Moroxydine hydrochloride | 3160-91-6 | MFCD00000075 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0420A5G
Encompass_Preferred
Moroxydine is a broad-spectrum antiviral agent with multi-antiviral activities against both DNA and RNA viruses, such as influenza, herpes simplex, and hepatitis C. It exhibits excellent antiviral activity and low cytotoxicity, effectively protecting cells from dsRNA and large DNA viruses like grass carp reovirus (GCRV) and giant salamander iridovirus (GSIV). It blocks virus-induced cytopathic effects, maintains normal cell morphology, and significantly inhibits apoptosis.
  • Demonstrates protective effects against GCRV and GSIV.
  • Blocks virus-induced cytopathic effects and cell death.
  • Decreases apoptosis and inhibits caspase 3 activity.
  • Down-regulates Bcl-2 levels and Bax expression.
  • Suppresses viral gene expression.
  • Arrests cell cycle in G1/S and G2/M phases.
  • Inhibits GCRV-induced upregulation of immune genes.
Catalog No. 50-004-08768