Learn More
Medchemexpress LLC Moroxydine hydrochloride (standard) | 3160-91-6 | MFCD00012798 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0420A1G
Encompass_Preferred
Moroxydine (ABOB) hydrochloride (Standard) is the analytical standard of Moroxydine (ABOB) hydrochloride. It is a broad-spectrum agent with multi-antiviral activities against DNA and RNA viruses. It exhibits excellent antiviral activity and low cytotoxicity to cells.
  • Multi-antiviral activities against DNA and RNA viruses
  • Effective against influenza virus, herpes simplex, varicella zoster, measles, mumps, and hepatitis C virus
  • Blocks GCRV-induced cytopathic effects
  • Eliminates nucleocapsids in CIK cells
  • Inhibits apoptosis, caspase 3 activity, Bax expression, and down-regulates Bcl-2 levels
  • Used as a reference standard for assay
  • Commonly used in HPLC, GC, and MS research experiments
Catalog No. 50-004-08767