Learn More
Medchemexpress LLC Morphiceptin | 74135-04-9 | 99.07% | 521.61 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP17015MG
Encompass_Preferred
Morphiceptin is a potent and specific agonist for morphine (μ) receptors. This synthetic peptide is the amide of a fragment of the milk protein β-casein. It exhibits morphinelike activities and is highly specific for morphine (μ) receptors.
  • Potent and specific agonist for morphine (μ) receptors
  • Synthetic peptide derived from β-casein
  • Exhibits morphinelike activities
  • Inhibits binding of [3H]dihydromorphine, 125I-labeled FK33824, and [3H]naloxone to rat brain membrane preparations with an IC50 of approximately 20 nM
Catalog No. 50-004-24272