Learn More
Medchemexpress LLC Morphiceptin | 74135-04-9 | 99.1% | 521.61 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP17011MG
Encompass_Preferred
Morphiceptin is a potent and specific agonist for morphine (μ) receptors. It is a synthetic peptide, derived from β-casein. It exhibits morphinelike activities and shows high specificity for morphine (μ) receptors, inhibiting the binding of [3H]dihydromorphine, 125I-labeled FK33824, and [3H]naloxone to rat brain membrane preparations with an IC50 of about 20 nM.
  • Potent and specific agonist for morphine (μ) receptors
  • Synthetic peptide derived from β-casein
  • White to off-white solid appearance
  • Soluble in water and DMSO
Catalog No. 50-004-23506