Learn More
Medchemexpress LLC Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)- | 880759-65-9 | 98.1% | 723.69 | C38H44Cl2N4O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1010115MG
Encompass_Preferred
Naloxonazine dihydrochloride is the dihydrochloride salt of naloxonazine, a selective μ-opioid receptor antagonist used as a research reagent. It exhibits potent MOR antagonism (IC50 ≈ 5.4 nM) and has shown anti-leishmanial activity in cell-based assays. The material is supplied as a solid powder for biochemical and pharmacological studies.
  • Selective μ-opioid receptor antagonist with high potency.
  • Shows anti-leishmanial activity in vitro against intracellular amastigotes.
  • Supplied as a solid powder suitable for formulation and dosing in assays.
  • Available in small pack sizes for laboratory experiments.
  • Characterized with defined molecular weight and composition for reproducibility.
Catalog No. 50-002-12058