Learn More
Medchemexpress LLC Morpholine,4-[(4-bromophenyl)methyl]- | 132833-51-3 | 99.9% | C11H14BrNO | 1 G

Supplier: Medchemexpress LLC HY768771G
4-(4-Bromobenzyl)morpholine, also known as Morpholine,4-[(4-bromophenyl)methyl]-, is a chemical compound that appears as a white to off-white solid. It exhibits a high purity of 99.94% (HPLC) and is intended for research use only.
- Appears as a white to off-white solid
- High purity of 99.94% (HPLC)
- Suitable for research applications
- Stable for 3 years at room temperature
- Stable for 2 years at -80°C in solvent, or 1 year at -20°C in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.