Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Morpholine,4-[(4-bromophenyl)methyl]- | 132833-51-3 | 99.9% | C11H14BrNO | 100 G
SDP

Supplier:  Medchemexpress LLC HY76877100G

Encompass_Preferred

This is Morpholine,4-[(4-bromophenyl)methyl]-, a high-purity chemical compound. With a purity of 99.9%, it is suitable for various laboratory applications as a chemical intermediate.

  • High purity of 99.9%
  • Molecular formula C11H14BrNO
  • CAS number 132833-51-3
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-33971


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.