Learn More
Crescent Chemical Co Inc MORPHOLINOETHANE SULFONIC ACID
SDP
Supplier:  Crescent Chemical Co Inc SE29830.03
Encompass_Preferred
Morpholinoethane sulfonic acid, monohydrate analytical grade pKa 20 = 6.15. Buffering substance (1). Physical constants (2). Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 29830.03 CAS 145224-94-8
Catalog No. 50-247-882