Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue grinder with a radial serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm. Interchangeable parts. Unit is autoclavable.
SDP
Supplier:  Electron Microscopy Sciences 6479530
Encompass_Preferred
Potter-Elvehjem Tissue Grinder incorporates a serrated pestle design to disperse homogenate into the mortar cylinder. Standard unit with Teflon pestles are well suited for soft tissue, such as brain and liver. The tissue grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. This unit is autoclavable and can be used for power homogenization as with overhead stirrer. Depth of the serration is 0.20" to 0.25" (0.1 to 0.15 mm).
Catalog No. NC3049627